Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGCACTTGCCCTTTGGTAATCC[C/T]GCTCCTTCTGAATCTGTTCCACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608023 MIM: 612038 | ||||||||||||||||||||
Literature Links: |
DDX56 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX56 - DEAD-box helicase 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED4 - transmembrane p24 trafficking protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303058.1 | 610 | Missense Mutation | CAG,CGG | Q,R 174 | NP_001289987.1 | |
NM_001303059.1 | 610 | Intron | NP_001289988.1 | |||
NM_001303060.1 | 610 | Missense Mutation | CAG,CGG | Q,R 115 | NP_001289989.1 | |
NM_001303061.1 | 610 | Missense Mutation | CAG,CGG | Q,R 115 | NP_001289990.1 | |
NM_001303062.1 | 610 | Missense Mutation | CAG,CGG | Q,R 115 | NP_001289991.1 | |
NM_182547.3 | 610 | Missense Mutation | CAG,CGG | Q,R 174 | NP_872353.2 |