Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTCGAAGAGCACTTTGTGGCCT[C/T]CTCAGTGGGTGAGATGTGGCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613883 MIM: 606679 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf34 - chromosome 7 open reading frame 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178829.4 | 289 | Missense Mutation | TCC,TTC | S,F 83 | NP_849151.2 |
KEL - Kell blood group, metallo-endopeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000420.2 | 289 | Intron | NP_000411.1 | |||
XM_005249993.2 | 289 | Intron | XP_005250050.1 | |||
XM_005249994.3 | 289 | Intron | XP_005250051.1 |
TRPV5 - transient receptor potential cation channel subfamily V member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |