Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTGCTTGAGGCTGAAGGTCAC[A/G]CTGCTTGGTCTTCACTGCTGTATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609080 | ||||||||||||||||||||
Literature Links: |
FAM185A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM185A - family with sequence similarity 185 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145268.1 | 2435 | Intron | NP_001138740.1 | |||
NM_001145269.1 | 2435 | Intron | NP_001138741.1 | |||
XM_006715896.3 | 2435 | Intron | XP_006715959.1 | |||
XM_011515922.2 | 2435 | Intron | XP_011514224.1 | |||
XM_011515923.2 | 2435 | Intron | XP_011514225.1 | |||
XM_011515924.2 | 2435 | Intron | XP_011514226.1 | |||
XM_011515925.2 | 2435 | Intron | XP_011514227.1 | |||
XM_011515926.2 | 2435 | Intron | XP_011514228.1 | |||
XM_017011846.1 | 2435 | Intron | XP_016867335.1 | |||
XM_017011847.1 | 2435 | Intron | XP_016867336.1 | |||
XM_017011848.1 | 2435 | Intron | XP_016867337.1 | |||
XM_017011849.1 | 2435 | Intron | XP_016867338.1 |
FBXL13 - F-box and leucine rich repeat protein 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111038.1 | 2435 | Missense Mutation | GCG,GTG | A,V 690 | NP_001104508.1 | |
NM_001287150.1 | 2435 | Missense Mutation | GCG,GTG | A,V 707 | NP_001274079.1 | |
NM_145032.3 | 2435 | Missense Mutation | GCG,GTG | A,V 735 | NP_659469.3 | |
XM_005250205.3 | 2435 | Missense Mutation | GCG,GTG | A,V 825 | XP_005250262.1 | |
XM_005250207.3 | 2435 | Missense Mutation | GCG,GTG | A,V 792 | XP_005250264.1 | |
XM_005250208.3 | 2435 | Missense Mutation | GCG,GTG | A,V 780 | XP_005250265.1 | |
XM_005250209.2 | 2435 | Missense Mutation | GCG,GTG | A,V 735 | XP_005250266.1 | |
XM_006715898.2 | 2435 | Missense Mutation | GCG,GTG | A,V 584 | XP_006715961.1 | |
XM_011515928.2 | 2435 | Missense Mutation | GCG,GTG | A,V 797 | XP_011514230.1 | |
XM_011515929.2 | 2435 | Intron | XP_011514231.1 | |||
XM_011515930.2 | 2435 | Missense Mutation | GCG,GTG | A,V 735 | XP_011514232.1 | |
XM_011515932.2 | 2435 | Intron | XP_011514234.1 | |||
XM_017011850.1 | 2435 | Missense Mutation | GCG,GTG | A,V 822 | XP_016867339.1 | |
XM_017011851.1 | 2435 | Intron | XP_016867340.1 | |||
XM_017011852.1 | 2435 | Missense Mutation | GCG,GTG | A,V 780 | XP_016867341.1 | |
XM_017011853.1 | 2435 | Missense Mutation | GCG,GTG | A,V 735 | XP_016867342.1 |