Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGCCACTAGGACTGAAGTCCC[C/G]CATCAAGATCCAGCTTGGCAGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5J2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5J2 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003713.2 | 157 | Missense Mutation | CGG,GGG | R,G 27 | NP_001003713.1 | |
NM_001003714.2 | 157 | Missense Mutation | CGG,GGG | R,G 33 | NP_001003714.1 | |
NM_001039178.2 | 157 | Missense Mutation | CGG,GGG | R,G 27 | NP_001034267.1 | |
NM_004889.3 | 157 | Missense Mutation | CGG,GGG | R,G 33 | NP_004880.1 |
ATP5J2-PTCD1 - ATP5J2-PTCD1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198879.1 | 157 | Missense Mutation | CGG,GGG | R,G 27 | NP_001185808.1 |
CPSF4 - cleavage and polyadenylation specific factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |