Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAATGGCGGCGCCGAGCGCGGG[A/G]TCTTGGTCCACCTTCCAGCACAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600639 MIM: 616524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP2 - caspase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001224.4 | 262 | Intron | NP_001215.1 | |||
NM_032982.3 | 262 | Silent Mutation | GGA,GGG | G,G 7 | NP_116764.2 | |
NM_032983.3 | 262 | Silent Mutation | GGA,GGG | G,G 7 | NP_116765.2 |
LOC105375545 - uncharacterized LOC105375545 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM139 - transmembrane protein 139 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242773.2 | 262 | Intron | NP_001229702.1 | |||
NM_001242774.2 | 262 | Intron | NP_001229703.1 | |||
NM_001242775.2 | 262 | Intron | NP_001229704.1 | |||
NM_001282876.1 | 262 | Intron | NP_001269805.1 | |||
NM_001282877.1 | 262 | Intron | NP_001269806.1 | |||
NM_153345.3 | 262 | Intron | NP_699176.1 |