Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGAGAAACGGGGTGGTACATACT[C/G]GGTAAGTTCAGGGAAGGGCATGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610334 MIM: 610385 | ||||||||||||||||||||
Literature Links: |
TMEM176A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM176A - transmembrane protein 176A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018487.2 | 554 | Missense Mutation | TCG,TGG | S,W 114 | NP_060957.2 | |
XM_011516376.2 | 554 | Missense Mutation | TCG,TGG | S,W 131 | XP_011514678.1 | |
XM_011516377.2 | 554 | Missense Mutation | TCG,TGG | S,W 131 | XP_011514679.1 | |
XM_011516378.2 | 554 | Missense Mutation | TCG,TGG | S,W 131 | XP_011514680.1 | |
XM_017012393.1 | 554 | Missense Mutation | TCG,TGG | S,W 114 | XP_016867882.1 |
TMEM176B - transmembrane protein 176B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |