Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTTCTCCGCATGTGGGACTCT[C/T]GGCCTCATTATGAGTGGCATCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605215 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf71 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf71 - chromosome 7 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145531.2 | 3500 | Silent Mutation | CTC,CTT | L,L 98 | NP_001139003.1 | |
NM_001302625.1 | 3500 | Silent Mutation | CTC,CTT | L,L 73 | NP_001289554.1 | |
XM_017012010.1 | 3500 | Silent Mutation | CTC,CTT | L,L 98 | XP_016867499.1 | |
XM_017012011.1 | 3500 | Silent Mutation | CTC,CTT | L,L 98 | XP_016867500.1 | |
XM_017012012.1 | 3500 | Silent Mutation | CTC,CTT | L,L 95 | XP_016867501.1 | |
XM_017012013.1 | 3500 | Silent Mutation | CTC,CTT | L,L 66 | XP_016867502.1 | |
XM_017012014.1 | 3500 | Silent Mutation | CTC,CTT | L,L 73 | XP_016867503.1 |
SKAP2 - src kinase associated phosphoprotein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |