Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCGTGCTCACATTGAGGTAGGC[C/T]GAGGCGTTGGAGTTGCCTGCAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610486 MIM: 602181 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC4 - leucine rich repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022143.4 | 1751 | Silent Mutation | NP_071426.1 | |||
XM_011516461.2 | 1751 | Silent Mutation | XP_011514763.1 | |||
XM_017012505.1 | 1751 | Silent Mutation | XP_016867994.1 |
SND1 - staphylococcal nuclease and tudor domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017011987.1 | 1751 | Intron | XP_016867476.1 |