Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTCTGGGAAGGGCTGAGGCCTG[C/G]AGCCAGTGCCTGTTTCCCACTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf49 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf49 - chromosome 7 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243749.1 | 671 | UTR 3 | NP_001230678.1 | |||
NM_001243751.1 | 671 | UTR 3 | NP_001230680.1 | |||
NM_001243752.1 | 671 | UTR 3 | NP_001230681.1 | |||
NM_001243753.1 | 671 | UTR 3 | NP_001230682.1 | |||
NM_001243754.1 | 671 | Missense Mutation | CCA,GCA | P,A 65 | NP_001230683.1 | |
NM_001243755.1 | 671 | Missense Mutation | CCA,GCA | P,A 65 | NP_001230684.1 | |
NM_001305629.1 | 671 | Missense Mutation | CCA,GCA | P,A 107 | NP_001292558.1 | |
NM_001305630.1 | 671 | Intron | NP_001292559.1 | |||
NM_024033.3 | 671 | Missense Mutation | CCA,GCA | P,A 120 | NP_076938.2 | |
XM_017012589.1 | 671 | Missense Mutation | CCA,GCA | P,A 120 | XP_016868078.1 | |
XM_017012590.1 | 671 | Missense Mutation | CCA,GCA | P,A 120 | XP_016868079.1 | |
XM_017012591.1 | 671 | Missense Mutation | CCA,GCA | P,A 120 | XP_016868080.1 | |
XM_017012592.1 | 671 | Missense Mutation | CCA,GCA | P,A 107 | XP_016868081.1 | |
XM_017012593.1 | 671 | Missense Mutation | CCA,GCA | P,A 65 | XP_016868082.1 | |
XM_017012594.1 | 671 | Missense Mutation | CCA,GCA | P,A 65 | XP_016868083.1 | |
XM_017012595.1 | 671 | Intron | XP_016868084.1 |
TMEM140 - transmembrane protein 140 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018295.4 | 671 | Intron | NP_060765.4 |