Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTGCCACCCGGTTGGGGTTCT[C/T]GATGTCGATGAGCCCTTCAACGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604223 MIM: 603477 MIM: 607075 | ||||||||||||||||||||
Literature Links: |
ARPC1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARPC1B - actin related protein 2/3 complex subunit 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BUD31 - BUD31 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDAP1 - PDGFA associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014891.6 | 384 | Missense Mutation | AAG,GAG | K,E 84 | NP_055706.1 |