Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGTGCAGACAGAACATGAGCGA[G/T]TCTGGCTTCGTGACTACCGACCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EGFR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EGFR - epidermal growth factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005228.3 | Intron | NP_005219.2 | ||||
NM_201282.1 | Intron | NP_958439.1 | ||||
NM_201283.1 | Intron | NP_958440.1 | ||||
NM_201284.1 | Intron | NP_958441.1 |