Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGATTTTACTCCACAGTACGGTA[A/G]ATTGTGGGCTAGAATCTAAAAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATXN7L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATXN7L1 - ataxin 7 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDHR3 - cadherin related family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301161.1 | Intron | NP_001288090.1 | ||||
NM_152750.4 | Intron | NP_689963.2 | ||||
XM_005250224.4 | Intron | XP_005250281.1 | ||||
XM_005250225.2 | Intron | XP_005250282.1 | ||||
XM_006715905.2 | Intron | XP_006715968.1 | ||||
XM_011515955.2 | Intron | XP_011514257.1 | ||||
XM_011515956.2 | Intron | XP_011514258.1 | ||||
XM_011515957.2 | Intron | XP_011514259.1 | ||||
XM_011515958.2 | Intron | XP_011514260.1 | ||||
XM_011515959.2 | Intron | XP_011514261.1 | ||||
XM_017011862.1 | Intron | XP_016867351.1 | ||||
XM_017011863.1 | Intron | XP_016867352.1 | ||||
XM_017011864.1 | Intron | XP_016867353.1 | ||||
XM_017011865.1 | Intron | XP_016867354.1 | ||||
XM_017011866.1 | Intron | XP_016867355.1 | ||||
XM_017011867.1 | Intron | XP_016867356.1 |