Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTGTCTCTTGACCAGCTGCGA[A/G]CAGAAGCGCTGCAGCAGCCCGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615783 MIM: 602186 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAT16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAT16 - N-acetyltransferase 16 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198571.2 | 668 | Silent Mutation | TGC,TGT | C,C 143 | NP_940973.2 | |
XM_005250325.1 | 668 | Silent Mutation | TGC,TGT | C,C 143 | XP_005250382.1 | |
XM_011516168.1 | 668 | Silent Mutation | TGC,TGT | C,C 143 | XP_011514470.1 |
VGF - VGF nerve growth factor inducible | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |