Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGGCGCAGCTGTGGATAGCCTC[A/G]CCGCAATGATAGTAATTTATGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610129 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHST12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHST12 - carbohydrate (chondroitin 4) sulfotransferase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243794.1 | Intron | NP_001230723.1 | ||||
NM_001243795.1 | Intron | NP_001230724.1 | ||||
NM_018641.4 | Intron | NP_061111.1 | ||||
XM_011515443.2 | Intron | XP_011513745.1 | ||||
XM_011515444.2 | Intron | XP_011513746.1 |