Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTACAGTTCATTTTCAGTCATCA[A/G]ATAACATACATCACTTCCAAAACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608798 MIM: 606959 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DFNA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DFNA5 - DFNA5, deafness associated tumor suppressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127453.1 | 2272 | UTR 3 | NP_001120925.1 | |||
NM_001127454.1 | 2272 | UTR 3 | NP_001120926.1 | |||
NM_004403.2 | 2272 | UTR 3 | NP_004394.1 | |||
XM_017011802.1 | 2272 | UTR 3 | XP_016867291.1 |
MPP6 - membrane palmitoylated protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |