Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGGGAAGAGCGCCACTGGGAA[C/T]AGCATCCTGGGCCAGAGACGGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608084 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIMAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIMAP1 - GTPase, IMAP family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130759.3 | 275 | Silent Mutation | AAC,AAT | N,N 45 | NP_570115.1 |
GIMAP1-GIMAP5 - GIMAP1-GIMAP5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199577.1 | 275 | Silent Mutation | AAC,AAT | N,N 45 | NP_001186506.1 | |
NM_001303630.1 | 275 | Intron | NP_001290559.1 |