Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTGCCTCAGGGCCTGGAAGAGGC[C/T]AACGAGGGACTGCGCCCAGATGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604309 | ||||||||||||||||||||
Literature Links: |
FAM180A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM180A - family with sequence similarity 180 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_205855.3 | 726 | Missense Mutation | AGC,GGC | S,G 154 | NP_995327.1 | |
XM_006715983.3 | 726 | Missense Mutation | AGC,GGC | S,G 154 | XP_006716046.1 |
SLC13A4 - solute carrier family 13 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |