Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAATAAAAGGATGCAGCAACAA[C/T]GGAATGTAAGTTGTTGGATTAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606589 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNX13 - sorting nexin 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015132.4 | 5950 | UTR 3 | NP_055947.1 | |||
XM_005249672.1 | 5950 | UTR 3 | XP_005249729.1 | |||
XM_005249673.4 | 5950 | Intron | XP_005249730.1 | |||
XM_011515229.1 | 5950 | UTR 3 | XP_011513531.1 | |||
XM_011515230.2 | 5950 | Intron | XP_011513532.1 | |||
XM_011515231.2 | 5950 | Intron | XP_011513533.1 | |||
XM_011515233.2 | 5950 | Intron | XP_011513535.1 | |||
XM_017011893.1 | 5950 | UTR 3 | XP_016867382.1 | |||
XM_017011894.1 | 5950 | Intron | XP_016867383.1 | |||
XM_017011895.1 | 5950 | Intron | XP_016867384.1 | |||
XM_017011896.1 | 5950 | Intron | XP_016867385.1 | |||
XM_017011897.1 | 5950 | UTR 3 | XP_016867386.1 |