Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCAGCAGGATGTAGTGGACCTC[A/G]TGGTAGCGTGGATCCGAGGGGGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615732 MIM: 612548 | ||||||||||||||||||||
Literature Links: |
NSUN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NSUN5 - NOP2/Sun RNA methyltransferase family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168347.2 | Intron | NP_001161819.1 | ||||
NM_001168348.2 | Intron | NP_001161820.1 | ||||
NM_018044.4 | Intron | NP_060514.1 | ||||
NM_148956.3 | Intron | NP_683759.1 | ||||
XM_005277604.4 | Intron | XP_005277661.1 |
TRIM50 - tripartite motif containing 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |