Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGATTCCAGTGGGACCGGGCAGAC[A/G]CTGGATTCTTCCCCAGTGGCTTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608595 MIM: 608596 | ||||||||||||||||||||
Literature Links: |
NPSR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPSR1 - neuropeptide S receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300933.1 | 228 | Silent Mutation | ACA,ACG | T,T 18 | NP_001287862.1 | |
NM_001300934.1 | 228 | Silent Mutation | ACA,ACG | T,T 18 | NP_001287863.1 | |
NM_001300935.1 | 228 | Silent Mutation | ACA,ACG | T,T 18 | NP_001287864.1 | |
NM_207172.1 | 228 | Silent Mutation | ACA,ACG | T,T 18 | NP_997055.1 | |
NM_207173.1 | 228 | Silent Mutation | ACA,ACG | T,T 18 | NP_997056.1 |
NPSR1-AS1 - NPSR1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |