Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTGGTCAATTGTAGGTAGATA[A/T]GAAATAGCCAAGTGGGGCCTTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606246 MIM: 605326 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAZF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JAZF1 - JAZF zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175061.3 | 2445 | UTR 3 | NP_778231.2 | |||
XM_006715656.1 | 2445 | UTR 3 | XP_006715719.1 | |||
XM_017011831.1 | 2445 | UTR 3 | XP_016867320.1 | |||
XM_017011832.1 | 2445 | UTR 3 | XP_016867321.1 |
TAX1BP1 - Tax1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |