Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCAGGGGGATCCCAAAGATGAA[C/T]TCGATCTCGTAGCCGTGGGGCACCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100740 MIM: 614469 MIM: 611481 | ||||||||||||||||||||
Literature Links: |
ACHE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACHE - acetylcholinesterase (Cartwright blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000665.4 | 1375 | Silent Mutation | GAA,GAG | E,E 483 | NP_000656.1 | |
NM_001282449.1 | 1375 | Silent Mutation | GAA,GAG | E,E 395 | NP_001269378.1 | |
NM_001302621.1 | 1375 | Silent Mutation | GAA,GAG | E,E 483 | NP_001289550.1 | |
NM_001302622.1 | 1375 | Silent Mutation | GAA,GAG | E,E 483 | NP_001289551.1 | |
NM_015831.2 | 1375 | Silent Mutation | GAA,GAG | E,E 483 | NP_056646.1 | |
XM_006715995.2 | 1375 | Silent Mutation | GAA,GAG | E,E 483 | XP_006716058.1 | |
XM_011516225.1 | 1375 | Silent Mutation | GAA,GAG | E,E 649 | XP_011514527.1 | |
XM_011516226.1 | 1375 | Silent Mutation | GAA,GAG | E,E 561 | XP_011514528.1 | |
XM_011516228.2 | 1375 | Silent Mutation | GAA,GAG | E,E 483 | XP_011514530.1 | |
XM_011516229.2 | 1375 | Silent Mutation | GAA,GAG | E,E 483 | XP_011514531.1 | |
XM_017012219.1 | 1375 | Silent Mutation | GAA,GAG | E,E 649 | XP_016867708.1 | |
XM_017012220.1 | 1375 | Silent Mutation | GAA,GAG | E,E 561 | XP_016867709.1 |
SRRT - serrate, RNA effector molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFSP1 - UFM1 specific peptidase 1 (inactive) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |