Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCTTGGCCAGGCTGTTGTACAC[A/G]GGCTCTAGCTGCTTGCAGTGCCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PDIA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDIA4 - protein disulfide isomerase family A member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004911.4 | 1971 | Silent Mutation | CCC,CCT | P,P 563 | NP_004902.1 | |
XM_006716185.2 | 1971 | Silent Mutation | CCC,CCT | P,P 564 | XP_006716248.1 |