Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTCAGTCCCGGCAGGAGTCCAC[A/G]CGGAGGCTCATCCTTGTTGGGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608084 | ||||||||||||||||||||
Literature Links: |
GIMAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIMAP1 - GTPase, IMAP family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130759.3 | 221 | Silent Mutation | ACA,ACG | T,T 27 | NP_570115.1 |
GIMAP1-GIMAP5 - GIMAP1-GIMAP5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199577.1 | 221 | Silent Mutation | ACA,ACG | T,T 27 | NP_001186506.1 | |
NM_001303630.1 | 221 | Intron | NP_001290559.1 |