Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTATTGCTTTACAGATGGCAGAA[A/G]ATTTTTTCATTTAATGTCTGGGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BCAP29 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCAP29 - B-cell receptor-associated protein 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008405.2 | 437 | Silent Mutation | AAA,AAG | K,K 34 | NP_001008405.1 | |
NM_018844.3 | 437 | Silent Mutation | AAA,AAG | K,K 34 | NP_061332.2 | |
XM_006716051.2 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_006716114.1 | |
XM_011516406.2 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_011514708.1 | |
XM_011516407.1 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_011514709.1 | |
XM_011516408.2 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_011514710.1 | |
XM_011516409.2 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_011514711.1 | |
XM_017012436.1 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_016867925.1 | |
XM_017012437.1 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_016867926.1 | |
XM_017012438.1 | 437 | Silent Mutation | AAA,AAG | K,K 34 | XP_016867927.1 |
DUS4L - dihydrouridine synthase 4 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |