Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGTATCTCAATTATAGAAATAA[A/T]AGGAATTATGGCCATAGAATTGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616071 | ||||||||||||||||||||
Literature Links: |
C7orf31 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C7orf31 - chromosome 7 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138811.3 | 1209 | Missense Mutation | TAT,TTT | Y,F 586 | NP_620166.3 | |
XM_005249619.3 | 1209 | Missense Mutation | TAT,TTT | Y,F 586 | XP_005249676.1 | |
XM_005249620.3 | 1209 | Missense Mutation | TAT,TTT | Y,F 586 | XP_005249677.1 | |
XM_011515125.2 | 1209 | Missense Mutation | TAT,TTT | Y,F 376 | XP_011513427.1 |