Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGATGCCTGAGGAGTGCGAGCT[C/G]CACAGCAACATGGCAGGCAGGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194510 MIM: 601261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF3 - zinc finger protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278284.1 | 1709 | Intron | NP_001265213.1 | |||
NM_001278287.1 | 1709 | Intron | NP_001265216.1 | |||
NM_001278290.1 | 1709 | Intron | NP_001265219.1 | |||
NM_001278291.1 | 1709 | Intron | NP_001265220.1 | |||
NM_001278292.1 | 1709 | Intron | NP_001265221.1 | |||
NM_001318135.1 | 1709 | Intron | NP_001305064.1 | |||
NM_001318136.1 | 1709 | Intron | NP_001305065.1 | |||
NM_001318137.1 | 1709 | UTR 3 | NP_001305066.1 | |||
NM_017715.3 | 1709 | UTR 3 | NP_060185.2 | |||
NM_032924.4 | 1709 | Intron | NP_116313.3 | |||
XM_006716111.1 | 1709 | Intron | XP_006716174.1 | |||
XM_017012582.1 | 1709 | Intron | XP_016868071.1 | |||
XM_017012583.1 | 1709 | Intron | XP_016868072.1 |
ZSCAN21 - zinc finger and SCAN domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145914.2 | 1709 | UTR 3 | NP_666019.1 | |||
XM_005250568.4 | 1709 | UTR 3 | XP_005250625.1 | |||
XM_006716113.3 | 1709 | UTR 3 | XP_006716176.1 | |||
XM_017012584.1 | 1709 | UTR 3 | XP_016868073.1 | |||
XM_017012585.1 | 1709 | UTR 3 | XP_016868074.1 |