Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCAAACAAGGTGGAAGCAGGGCA[A/G]ATATGATGAGGATGGCAAACCCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF394 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF394 - zinc finger protein 394 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032164.2 | 1025 | Intron | NP_115540.2 | |||
XM_017012710.1 | 1025 | UTR 3 | XP_016868199.1 | |||
XM_017012711.1 | 1025 | Intron | XP_016868200.1 |
ZNF789 - zinc finger protein 789 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013258.1 | 1025 | Intron | NP_001013276.1 | |||
NM_213603.2 | 1025 | Missense Mutation | AAA,AGA | K,R 174 | NP_998768.2 | |
XM_011516067.2 | 1025 | Missense Mutation | AAA,AGA | K,R 167 | XP_011514369.1 | |
XM_011516070.2 | 1025 | Missense Mutation | AAA,AGA | K,R 157 | XP_011514372.1 | |
XM_011516073.2 | 1025 | Missense Mutation | AAA,AGA | K,R 79 | XP_011514375.1 | |
XM_017012018.1 | 1025 | Missense Mutation | AAA,AGA | K,R 174 | XP_016867507.1 | |
XM_017012019.1 | 1025 | Missense Mutation | AAA,AGA | K,R 157 | XP_016867508.1 | |
XM_017012020.1 | 1025 | Missense Mutation | AAA,AGA | K,R 157 | XP_016867509.1 | |
XM_017012021.1 | 1025 | Missense Mutation | AAA,AGA | K,R 157 | XP_016867510.1 | |
XM_017012022.1 | 1025 | Missense Mutation | AAA,AGA | K,R 157 | XP_016867511.1 | |
XM_017012023.1 | 1025 | Missense Mutation | AAA,AGA | K,R 136 | XP_016867512.1 | |
XM_017012024.1 | 1025 | Missense Mutation | AAA,AGA | K,R 116 | XP_016867513.1 | |
XM_017012025.1 | 1025 | Missense Mutation | AAA,AGA | K,R 79 | XP_016867514.1 | |
XM_017012026.1 | 1025 | Missense Mutation | AAA,AGA | K,R 79 | XP_016867515.1 | |
XM_017012027.1 | 1025 | Missense Mutation | AAA,AGA | K,R 79 | XP_016867516.1 |