Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGTGCTGGACGAGAGGAAGGCG[C/T]TGATCCCCTGCAAGAGGTACAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606570 | ||||||||||||||||||||
Literature Links: |
EPDR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPDR1 - ependymin related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242946.1 | 632 | Silent Mutation | CTG,TTG | L,L 85 | NP_001229875.2 | |
NM_001242948.1 | 632 | Intron | NP_001229877.1 | |||
NM_017549.4 | 632 | Silent Mutation | CTG,TTG | L,L 85 | NP_060019.2 |
SFRP4 - secreted frizzled related protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |