Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTGATGGCAGATGTAAATTGG[A/G]ATGTCGCCAGCCTCCGCAGCAGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114131 | ||||||||||||||||||||
Literature Links: |
CALCR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CALCR - calcitonin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164737.1 | 1597 | Silent Mutation | ATC,ATT | I,I 472 | NP_001158209.1 | |
NM_001164738.1 | 1597 | Silent Mutation | ATC,ATT | I,I 438 | NP_001158210.1 | |
NM_001742.3 | 1597 | Silent Mutation | ATC,ATT | I,I 438 | NP_001733.1 |