Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCTCCGGGCCGTGGCCCACCT[A/G]CGGTCCCAGCTCTGGGCCCACCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605464 MIM: 612205 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCB8 - ATP binding cassette subfamily B member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282291.1 | 241 | Silent Mutation | CTA,CTG | L,L 66 | NP_001269220.1 | |
NM_001282292.1 | 241 | Silent Mutation | CTA,CTG | L,L 49 | NP_001269221.1 | |
NM_001282293.1 | 241 | Intron | NP_001269222.1 | |||
NM_007188.4 | 241 | Silent Mutation | CTA,CTG | L,L 49 | NP_009119.2 |
ATG9B - autophagy related 9B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |