Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGCAAAGAGTCATATTCCAGAA[C/T]TGGAGCAAACCCTGGATAATTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609987 | ||||||||||||||||||||
Literature Links: |
STRA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STRA8 - stimulated by retinoic acid 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182489.1 | 938 | Silent Mutation | CTG,TTG | L,L 100 | NP_872295.1 | |
XM_011516137.2 | 938 | Silent Mutation | CTG,TTG | L,L 78 | XP_011514439.1 | |
XM_011516138.2 | 938 | Silent Mutation | CTG,TTG | L,L 78 | XP_011514440.1 | |
XM_011516139.2 | 938 | Silent Mutation | CTG,TTG | L,L 78 | XP_011514441.1 | |
XM_017012150.1 | 938 | Silent Mutation | CTG,TTG | L,L 78 | XP_016867639.1 |