Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGGGGCCATCAGAAGCGATGCCG[C/T]GGCTGGGGGCCAAAGTGCATGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609948 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107986762 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107986762 - uncharacterized LOC107986762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF216 - ring finger protein 216 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207111.3 | 2755 | Silent Mutation | CCA,CCG | P,P 919 | NP_996994.1 | |
NM_207116.2 | 2755 | Silent Mutation | CCA,CCG | P,P 862 | NP_996999.1 | |
XM_005249785.2 | 2755 | Silent Mutation | CCA,CCG | P,P 919 | XP_005249842.1 | |
XM_011515436.1 | 2755 | Silent Mutation | CCA,CCG | P,P 484 | XP_011513738.1 | |
XM_017012363.1 | 2755 | Silent Mutation | CCA,CCG | P,P 862 | XP_016867852.1 | |
XM_017012364.1 | 2755 | Intron | XP_016867853.1 | |||
XM_017012365.1 | 2755 | Silent Mutation | CCA,CCG | P,P 484 | XP_016867854.1 |