Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACCAGCCCAAAAGAAAAATGGC[A/G]TTTGTTGCAACACAGGGGGCCACGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603015 | ||||||||||||||||||||
Literature Links: |
MIR3609 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR3609 - microRNA 3609 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA28 - small Cajal body-specific RNA 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRRAP - transformation/transcription domain associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244580.1 | 215 | Silent Mutation | GCA,GCG | A,A 2 | NP_001231509.1 | |
NM_003496.3 | 215 | Silent Mutation | GCA,GCG | A,A 2 | NP_003487.1 |