Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGACGTGCTCAGAGCTCCGATG[A/C]CAAGGCCAGGGACCATGGCGCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604609 MIM: 608424 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724094 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102724094 - uncharacterized LOC102724094 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUC12 - mucin 12, cell surface associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUC17 - mucin 17, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040105.1 | 57 | Missense Mutation | ACA,CCA | T,P 2 | NP_001035194.1 |