Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTAGGTGGCAAGGTCTCTGTAC[C/T]GAAGCCAGTCTGATTCCTTCACTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600418 | ||||||||||||||||||||
Literature Links: |
AMPH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMPH - amphiphysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001635.3 | 2559 | Missense Mutation | CAG,CGG | Q,R 675 | NP_001626.1 | |
NM_139316.2 | 2559 | Missense Mutation | CAG,CGG | Q,R 633 | NP_647477.1 | |
XM_006715689.3 | 2559 | Missense Mutation | CAG,CGG | Q,R 1205 | XP_006715752.1 | |
XM_006715690.3 | 2559 | Missense Mutation | CAG,CGG | Q,R 799 | XP_006715753.1 | |
XM_006715691.3 | 2559 | Intron | XP_006715754.1 | |||
XM_011515271.2 | 2559 | Missense Mutation | CAG,CGG | Q,R 841 | XP_011513573.1 | |
XM_017011995.1 | 2559 | Missense Mutation | CAG,CGG | Q,R 1247 | XP_016867484.1 | |
XM_017011996.1 | 2559 | Missense Mutation | CAG,CGG | Q,R 901 | XP_016867485.1 |
TRG-AS1 - T cell receptor gamma locus antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |