Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTTATCCTCCCTTCTCAAGCAT[A/G]CTCTGCAAGAAAAACAGAAAGAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613748 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHCHD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHCHD3 - coiled-coil-helix-coiled-coil-helix domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317177.1 | Intron | NP_001304106.1 | ||||
NM_001317178.1 | Intron | NP_001304107.1 | ||||
NM_017812.3 | Intron | NP_060282.1 |