Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGGTGGAGCGGAGTAAGATGAT[A/C]GACCGCAACCTCCGTGAGGACGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNAI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNAI1 - G protein subunit alpha i1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256414.1 | 394 | Intron | NP_001243343.1 | |||
NM_002069.5 | 394 | Silent Mutation | ATA,ATC | I,I 19 | NP_002060.4 |