Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCGGGAGCCTGTGTCAGGAAAGC[A/C]TGTCAGAGCAGAGCTGCCAGATGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616962 | ||||||||||||||||||||
Literature Links: |
GIMAP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIMAP8 - GTPase, IMAP family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175571.3 | 575 | Missense Mutation | ATG,CTG | M,L 1 | NP_783161.1 | |
XM_005249950.4 | 575 | Missense Mutation | ATG,CTG | M,L 1 | XP_005250007.1 | |
XM_005249951.3 | 575 | Missense Mutation | ATG,CTG | M,L 1 | XP_005250008.1 |