Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGCCCTGGCTTACTTGCCTTTC[C/T]TGTAGTTCTTTCAGCCGAAAGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616960 | ||||||||||||||||||||
Literature Links: |
GIMAP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIMAP6 - GTPase, IMAP family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244071.1 | 1003 | UTR 3 | NP_001231000.1 | |||
NM_001244072.1 | 1003 | Silent Mutation | CAA,CAG | Q,Q 319 | NP_001231001.1 | |
NM_024711.5 | 1003 | Silent Mutation | CAA,CAG | Q,Q 249 | NP_078987.3 |