Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAGGTAGGGATGCTCACTTTTA[C/T]CTTTCCGGCTCCTGTGAAATCCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCC1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCC1L - RCC1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281441.1 | 593 | Missense Mutation | GAT,GGT | D,G 150 | NP_001268370.1 | |
NM_030798.4 | 593 | Missense Mutation | GAT,GGT | D,G 150 | NP_110425.2 | |
NM_148842.2 | 593 | Missense Mutation | GAT,GGT | D,G 150 | NP_683682.1 |