Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCAAGATGGCCGTGGAGTATGA[C/T]CGCTTCATCGAGTCCGGCAGGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602576 | ||||||||||||||||||||
Literature Links: |
LFNG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LFNG - LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040167.1 | 575 | Silent Mutation | GAC,GAT | D,D 186 | NP_001035257.1 | |
NM_001040168.1 | 575 | Silent Mutation | GAC,GAT | D,D 186 | NP_001035258.1 | |
NM_001166355.1 | 575 | Silent Mutation | GAC,GAT | D,D 115 | NP_001159827.1 | |
NM_002304.2 | 575 | Silent Mutation | GAC,GAT | D,D 57 | NP_002295.1 |
MIR4648 - microRNA 4648 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |