Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGCTGTGTGGTCCCGCAAAGAG[A/G]CAGCTGAGCTTGGGCCTCAGGTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600312 MIM: 614905 | ||||||||||||||||||||
Literature Links: |
MIR6836 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6836 - microRNA 6836 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT1 - nudix hydrolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013321.3 | 1492 | Silent Mutation | TGC,TGT | C,C 442 | NP_037453.1 | |
XM_011515329.2 | 1492 | Silent Mutation | TGC,TGT | C,C 434 | XP_011513631.1 | |
XM_011515330.1 | 1492 | Silent Mutation | TGC,TGT | C,C 428 | XP_011513632.1 | |
XM_017012083.1 | 1492 | Silent Mutation | TGC,TGT | C,C 389 | XP_016867572.1 | |
XM_017012084.1 | 1492 | Silent Mutation | TGC,TGT | C,C 389 | XP_016867573.1 |