Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGTGGAGCTAGTTGAAGAGGAGT[C/T]CTCCTGTTCGTGCTAATTTAGAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL7 - kelch like family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031710.2 | 426 | Intron | NP_001026880.2 | |||
NM_001172428.1 | 426 | Intron | NP_001165899.1 | |||
NM_018846.4 | 426 | UTR 5 | NP_061334.4 | |||
XM_006715753.2 | 426 | Intron | XP_006715816.1 | |||
XM_006715754.2 | 426 | Intron | XP_006715817.1 | |||
XM_006715755.2 | 426 | UTR 5 | XP_006715818.1 | |||
XM_006715756.2 | 426 | UTR 5 | XP_006715819.1 | |||
XM_006715757.3 | 426 | Intron | XP_006715820.1 | |||
XM_017012439.1 | 426 | UTR 5 | XP_016867928.1 | |||
XM_017012440.1 | 426 | Intron | XP_016867929.1 | |||
XM_017012441.1 | 426 | Intron | XP_016867930.1 |
KLHL7-AS1 - KLHL7 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |