Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCATTTGCTTTCCTTTATTTGG[C/T]AAGGTACTTGACCATGTGATAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614048 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1324L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1324L - KIAA1324 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142749.2 | 3457 | UTR 3 | NP_001136221.1 | |||
NM_001291990.1 | 3457 | UTR 3 | NP_001278919.1 | |||
NM_001291991.1 | 3457 | UTR 3 | NP_001278920.1 | |||
NM_152748.3 | 3457 | UTR 3 | NP_689961.3 | |||
XM_006715894.3 | 3457 | UTR 3 | XP_006715957.1 | |||
XM_011515918.2 | 3457 | UTR 3 | XP_011514220.1 | |||
XM_011515921.1 | 3457 | UTR 3 | XP_011514223.1 | |||
XM_017011843.1 | 3457 | UTR 3 | XP_016867332.1 | |||
XM_017011844.1 | 3457 | UTR 3 | XP_016867333.1 |