Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACTCACCTGTAGTCTGCTGTTG[A/G]GGAATCTGAGTCTGATGTGGCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611686 MIM: 604149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASD1 - CAS1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022900.4 | 1448 | Intron | NP_075051.4 | |||
XM_006716092.3 | 1448 | Intron | XP_006716155.1 | |||
XM_006716093.3 | 1448 | Intron | XP_006716156.2 | |||
XM_011516495.2 | 1448 | Intron | XP_011514797.1 | |||
XM_017012546.1 | 1448 | Intron | XP_016868035.1 | |||
XM_017012547.1 | 1448 | Intron | XP_016868036.1 |