Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACGCAGCAATTTTTACTAAAAT[A/G]TGCCTGCTGAATGGTCAGAAAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608444 MIM: 602980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KMT2E - lysine methyltransferase 2E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRPK2 - SRSF protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278273.1 | Intron | NP_001265202.1 | ||||
NM_182691.2 | Intron | NP_872633.1 | ||||
NM_182692.2 | Intron | NP_872634.1 | ||||
XM_005250549.3 | Intron | XP_005250606.2 | ||||
XM_005250550.3 | Intron | XP_005250607.1 | ||||
XM_005250551.4 | Intron | XP_005250608.1 | ||||
XM_006716098.2 | Intron | XP_006716161.1 | ||||
XM_011516536.2 | Intron | XP_011514838.1 | ||||
XM_011516538.2 | Intron | XP_011514840.1 | ||||
XM_017012560.1 | Intron | XP_016868049.1 | ||||
XM_017012561.1 | Intron | XP_016868050.1 | ||||
XM_017012562.1 | Intron | XP_016868051.1 | ||||
XM_017012563.1 | Intron | XP_016868052.1 | ||||
XM_017012564.1 | Intron | XP_016868053.1 | ||||
XM_017012565.1 | Intron | XP_016868054.1 | ||||
XM_017012566.1 | Intron | XP_016868055.1 |