Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGGACTCGCTGTGCAGGGTCC[G/T]GCTCCGAGGCGCTGGTCGGCAGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142996 | ||||||||||||||||||||
Literature Links: |
EVX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EVX1 - even-skipped homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304519.1 | 919 | UTR 5 | NP_001291448.1 | |||
NM_001304520.1 | 919 | UTR 5 | NP_001291449.1 | |||
NM_001989.4 | 919 | Missense Mutation | GGC,TGC | G,C 145 | NP_001980.1 |
EVX1-AS - EVX1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |