Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGGGGAGCTGAAGTACCACCC[C/T]GAGTGTTTCATCTGCCTCACGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601329 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIMK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIMK1 - LIM domain kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204426.1 | 310 | Silent Mutation | CCC,CCT | P,P 72 | NP_001191355.1 | |
NM_002314.3 | 310 | Silent Mutation | CCC,CCT | P,P 106 | NP_002305.1 |