Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCTTGGCCGCCAGCAGGCGCA[A/G]GTTGCTTTCGCTAGGCCGGTAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615783 MIM: 602186 | ||||||||||||||||||||
Literature Links: |
NAT16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAT16 - N-acetyltransferase 16 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198571.2 | 1005 | Silent Mutation | CTG,TTG | L,L 256 | NP_940973.2 | |
XM_005250325.1 | 1005 | Silent Mutation | CTG,TTG | L,L 256 | XP_005250382.1 | |
XM_011516168.1 | 1005 | Silent Mutation | CTG,TTG | L,L 256 | XP_011514470.1 |
VGF - VGF nerve growth factor inducible | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |